Skip to Content
Merck
All Photos(2)

Documents

MABE1128

Sigma-Aldrich

Anti-SETBP1 Antibody, clone 63

clone 63, from mouse

Synonym(s):

SET-binding protein, SEB

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

63, monoclonal

species reactivity

human, mouse

technique(s)

western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... SETBP1(26040)

General description

SET-binding protein (UniProt Q9Y6X0; also known as SEB, SETBP1) is encoded by the SETBP1 (also known as KIAA0437, MRD29) gene (Gene ID 26040) in human. SETBP1 is shown to bind the SET nuclear oncogene involved in DNA replication. It is expressed in various tissues with low levels in myeloid and monocytic cells and in CD34+ cells, however, higher expression is observed in myeloid malignancies. Overexpression of SETBP1 promotes leukemogenesis by enhancing full-length SET protein and then impairing the phosphatase activity of the tumor suppressor PP2A through the formation of a SETBP1-SET-PP2A complex. Patients with SETBP1 overexpression have a significantly shorter overall survival, and poor prognosis, particularly in the elderly patients. Mutations in SETBP1 gene are associated with Schinzel-Giedion midface retraction syndrome, which is characterized by severe mental retardation, distinctive facial features, and multiple congenital malformations. (Ref.: Cristobal, I., et al. (2010). Blood 115(3): 615-625).


Specificity

Clone 63 targets the N-terminal region of human and mouse SETBP1.

Immunogen

GST-tagged recombinant human SETBP1 N-terminal fragment.

Application

Anti-SETBP1, clone 63, Cat. No. MABE1128, is a highly specific mouse monoclonal antibody that targets SETBP1 and has been tested in Western Blotting.
Research Category
Epigenetics & Nuclear Function
Western Blotting Analysis: A 1:4 dilution of clone 63 hybridoma culture supernatant detected SETBP1 in 30-50 µg of REH human acute lymphoid leukemia cell lysate (Courtesy of Jesper Christensen, Ph.D. and Kristian Helin, Ph.D., University of Copenhagen, Denmark).

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected SETBP1 in 10 µg of HeLa cell lysate.

Target description

~175 kDa observed. 175.0 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified.
Purified mouse IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service