Skip to Content
Merck
All Photos(1)

Key Documents

MABE1073

Sigma-Aldrich

Anti-acetyl SMC3 Antibody (Lys105/106), clone 21A7

clone 21A7, from mouse

Synonym(s):

Structural maintenance of chromosomes protein 3, Lys105/106 acetylated, Bamacan, Lys105/106 acetylated, Basement membrane-associated chondroitin proteoglycan, Lys105/106 acetylated, Chondroitin sulfate proteoglycan 6, Lys105/106 acetylated, Chromosome-as

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

21A7, monoclonal

species reactivity

human

species reactivity (predicted by homology)

nonhuman primates (based on 100% sequence homology), Xenopus (based on 100% sequence homology), bovine (based on 100% sequence homology), mouse (based on 100% sequence homology), rat (based on 100% sequence homology)

technique(s)

ChIP: suitable (ChIP-seq)
ELISA: suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

acetylation (Lys105/Lys106)

Gene Information

human ... SMC3(9126)

General description

Structural maintenance of chromosomes protein 3 (UniProt Q9UQE7; also known as Bamacan, Basement membrane-associated chondroitin proteoglycan, Chondroitin sulfate proteoglycan 6, Chromosome-associated polypeptide, hCAP, SMC protein 3, SMC-3) is encoded by the SMC3 (also known as BAM, BMH, CDLS3, CSPG6, SMC3L1) gene (Gene ID 9126) in human. Cohesion is a nuclear tripartite complex composed of Rad21 and two structural maintenance of chromosomes proteins, SMC1 and SMC3. Cohesin plays an important role in sister chromatid cohesion, as well as in DNA repair and gene expression regulation. To establish sister chromatid cohesion during S phase, SMC3 is acetylated (on K105 and K106 in human) by two cohesin acetyltransferases (CoATs), ESCO1 and ESCO2, that differ in their N-terminal domains and expression during development and across the cell cycle. ChIP-seq analysis reveals that ESCO1 is recruited by cohesin to over 11,000 chromatin sites occupied by cohesin and CTCF that mediate long-range chromatin interactions and regulate transcription globally. On ther other hand, ESCO2 is infrequently enriched at sites targeted by REST/NRSF (RE1-silencing transcription factor/neuron-restrictive silencer factor) that represses transcription of neuron-specific genes. SMC3 acetylation is thought to induce conformational changes of the dimeric hinge structure during S phase and to allow dimer opening and loading of DNA.

Specificity

Clone 21A7 bound Lys105/106-diacetylated SMC3 peptide with 2-fold higher affinity than Lys105-monoacetylated SMC3 peptide (Kd = 0.04 and 0.09 µg/mL, respectively), while displaying much reduced affinity toward Lys105-monoacetylated (Kd = 0.73 µg/mL) and little affinity toward nonacetylated (Kd = 2.99 µg/mL) peptide (Rahman, S., et al. (2015). Proc. Natl. Acad. Sci. U. S. A. 112(36):11270-11275).

Immunogen

KLH-conjugated linear peptide corresponding to the target region sequence of human/mouse/rat SMC3 containing acetylated Lys105 and Lys106.

Application

Anti-acetyl SMC3 (Lys105/106), clone 21A7, Cat. No. MABE1073, is a highly specific mouse monoclonal antibody that targets SMC3 Lys105/106 acetylation and has been tested in Chromatin Immunoprecipitation (ChIP), ChIP-seq, ELISA, and Western Blotting.
ELISA Analysis: A representative lot bound Lys105/106-diacetylated SMC3 peptide with 2-fold higher affinity than Lys105-monoacetylated SMC3 peptide (Kd = 0.04 and 0.09 µg/mL, respectively), while displaying much reduced affinity toward Lys105-monoacetylated (Kd = 0.73 µg/mL) and little affinity toward nonacetylated (Kd = 2.99 µg/mL) peptide (Rahman, S., et al. (2015). Proc. Natl. Acad. Sci. U. S. A. 112(36):11270-11275).

Chromatin Immunoprecipitation (ChIP) Analysis: A representative lot detected enhanced SMC3 occupancy at target chromatin sites in HDAC8-null HCT116 cells than wild-type HCT-116 cells (Rahman, S., et al. (2015). Proc. Natl. Acad. Sci. U. S. A. 112(36):11270-11275).

ChIP-seq Analysis: A representative lot detected SMC3 chromosome 11 enrichment sites, including 81% of Rad21 target sites and 70% of Esco1 target sites, by ChIP-seq (Rahman, S., et al. (2015). Proc. Natl. Acad. Sci. U. S. A. 112(36):11270-11275).

Western Blotting Analysis: A representative lot detected SMC3 Lys105/106 acetylation in HeLa cells. ESCO1 or ESCO2 depletion by shRNA treatment reduced SMC3 Lys105/106 acetylation level, while dual ESCO1/2 depletion resulted in most profound SMC3 Lys105/106 acetylation reduction (Rahman, S., et al. (2015). Proc. Natl. Acad. Sci. U. S. A. 112(36):11270-11275).
Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Western Blotting in human retinal epithelial pigment (REP) cells.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected a reduced SMC3 Lys105/106 acetylation in 10 µg of lysate from N-acetyltransferase ESCO1-knockout human retinal epithelial pigment (REP) cells than in lysate from wild-type REP cells.

Target description

~160 kDa observed. 142.0/141.5/141.6/138.4/140.7 kDa (bovine/human/mouse/rat/xenopus) calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified.
Purified mouse IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicole L Arruda et al.
Epigenetics & chromatin, 15(1), 30-30 (2022-08-20)
Cohesin is an important structural regulator of the genome, regulating both three-dimensional genome organization and gene expression. The core cohesin trimer interacts with various HEAT repeat accessory subunits, yielding cohesin complexes of distinct compositions and potentially distinct functions. The roles
Chunlong Zhao et al.
Journal of medicinal chemistry, 67(16), 14016-14039 (2024-08-02)
HDAC8 can mediate signals by using its enzymatic or nonenzymatic functions, which are expected to be critical for various types of cancer. Herein, we employed proteolysis targeting chimera (PROTAC) technology to target the enzymatic as well as the nonenzymatic functions
Tasha B Toro et al.
PloS one, 18(9), e0291779-e0291779 (2023-09-18)
Acetylation of lysine residues is an important and common post-translational regulatory mechanism occurring on thousands of non-histone proteins. Lysine deacetylases (KDACs or HDACs) are a family of enzymes responsible for removing acetylation. To identify the biological mechanisms regulated by individual
Jiaxin Zhang et al.
Nucleic acids research, 51(10), 4760-4773 (2023-03-14)
Besides entrapping sister chromatids, cohesin drives other high-order chromosomal structural dynamics like looping, compartmentalization and condensation. ESCO2 acetylates a subset of cohesin so that cohesion must be established and only be established between nascent sister chromatids. How this process is
Ayad A Al-Hamashi et al.
Bioorganic chemistry, 116, 105297-105297 (2021-09-13)
Despite the advances in treatment strategies, cancer is still the second leading cause of death in the USA. A majority of the currently used cancer drugs have limitations in their clinical use due to poor selectivity, toxic side effects and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service