Skip to Content
Merck
All Photos(1)

Key Documents

MABC1131

Sigma-Aldrich

Anti-PD-L1 B7-H1 Antibody, clone 10B5

clone 10B5, from hamster(Armenian)

Synonym(s):

Programmed cell death 1 ligand 1, PDCD1 ligand 1, Programmed death ligand 1, B7 homolog 1, B7-H1, CD274

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

biological source

hamster (Armenian)

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

10B5, monoclonal

species reactivity

mouse

packaging

antibody small pack of 25 μg

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable (paraffin)

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

mouse ... Cd274(60533)

Related Categories

General description

Programmed cell death 1 ligand 1 (UniProt: Q9EP73; also known as PD-L1, PDCD1 ligand 1, Programmed death ligand 1, B7 homolog 1, B7-H1, CD274) is encoded by the Cd274 (also known as B7h1, Pdcd1l1, Pdcd1lg1, Pdl1) gene (Gene ID: 50533) in murine species. PD-L1 is a single-pass type I membrane protein that is highly expressed in the heart, thymus, skeletal muscle, and lung. Its expression is also reported on activated dendritic cells, B-cells, and macrophages. Numerous tumor cell lines of lymphoid origin also express PD-L1. PD-L1 expression can be up-regulated by interferon-gamma treatment in monocytes, B cells, and macrophages. PD-1 and its ligands, PD-L1 and PD-L2 play a key role in the maintenance of peripheral tolerance, a process by which the quiescence of autoreactive mature T cells is maintained. However, tumors and pathogens that cause chronic infections can exploit this pathway to escape T-cell mediated tumor-specific and pathogen-specific immunity. The effector functions of T-cells expressing PD-1 can be downregulated by PD-L1 or PD-L2 expressed by the tumor cells. PD-L1 is initially produced with signal peptide (aa. 1-18) sequence, the removal of which yields the mature protein with a large extracellular (aa. 19-239) domain, a transmembrane domain (aa 240-260), and a cytoplasmic domain (aa 261-290).

Specificity

This Armenian hamster monoclonal antibody detects Programmed cell death 1 ligand 1 (PD-L1) in murine species.

Immunogen

Epitope: extracellular domain
Recombinant fragment corresponding to the extracellular domain of Mouse Programmed cell death 1 ligand 1.

Application

Anti-PD-L1 B7-H1, clone 10B5, Cat. No. MABC1131, is an Armenian hamster monoclonal antibody that detects Programmed cell death 1 ligand 1 (PD-L1) and has been tested for use in Flow Cytometry, Immunohistochemistry (Paraffin), and Inhibition Function assay.
Inhibits Activity/Function Analysis: A representative lot blocked the activity of PD-L1 B7-H1 and inhibited apoptosis of activated alloreactive 2C T cells. (Hirano, F., et. al. (2005). Cancer Res. 65(3):1089-96; Goldberg, M.V., et. al. (2007). Blood. 110(1):186-92; Joshi, T., et. al. (2009). PLoS Pathog. 5(5):e1000431; Liu, X., et. al. (2016). Sci Rep. 6:36722).



Flow Cytometry Analysis: A representative lot detected PD-L1 B7-H1 in Flow Cytometry applications (Hirano, F., et. al. (2005). Cancer Res. 65(3):1089-96; Dong, H., et. al. (2004). Immunity. 20(3):327-36).
Research Category
Apoptosis & Cancer

Quality

Evaluated by Immunohistochemistry (Paraffin) in mouse heart tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:25 dilution of this antibody detected PD-L1 B7-H1 in mouse heart tissue sections.

Target description

32.78 kDa calculated.

Physical form

Format: Purified
Protein G purified
Purified armenian hamster monoclonal antibody IgG2-IgG3 in PBS without azide.

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service