Skip to Content
Merck
All Photos(1)

Documents

MAB2499

Sigma-Aldrich

Anti-Fibrillin-1 Antibody, CT, clone 69

clone 69, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

69, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FBN1(2200)

Specificity

Monoclonal antibody MAB2499 recognizes human Fibrillin-1. Epitope mapping studies identify the binding site of this antibody to the C-terminal end of the molecule, between amino acid residues 2093 and 2871. The antibody is reactive with human, chicken, and bovine Fibrillin-1.

Immunogen

Epitope: C-terminus
Human Fibrillin-1

Application

Detect Fibrillin-1 using this Anti-Fibrillin-1 Antibody, C-terminus, clone 69 validated for use in ELISA, IP, WB & IC.
Immunoblotting

Immunofluorescence

Immunoprecipitation

ELISA

Optimal working dilutions must be determined by end user.
Research Category
Cell Structure
Research Sub Category
ECM Proteins

Physical form

Format: Purified
Purified immunoglobulin. Liquid at 1 mg/mL in 20 mM phosphate buffer, 250 mM NaCl, pH 7.6, containing 0.1% sodium azide.Note: Sodium azide is toxic. MSDS available upon request.

Storage and Stability

Maintain refrigerated at 2-8°C in undiluted aliquots for up to 12 months.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fan Chen et al.
Skin research and technology : official journal of International Society for Bioengineering and the Skin (ISBS) [and] International Society for Digital Imaging of Skin (ISDIS) [and] International Society for Skin Imaging (ISSI), 29(6), e13350-e13350 (2023-06-26)
Intrinsic skin aging is an inevitable process with reduced extracellular matrix deposition and impaired mechanical integrity in the dermal-epidermal junction (DEJ). Hyaluronan is one of the most promising natural ingredients. In this research, multiple mechanisms of a novel hyaluronan complex
Enrico Almici et al.
Frontiers in bioengineering and biotechnology, 10, 851825-851825 (2022-05-14)
Collagen VI-related dystrophies (COL6-RDs) are a group of rare congenital neuromuscular dystrophies that represent a continuum of overlapping clinical phenotypes that go from the milder Bethlem myopathy (BM) to the severe Ullrich congenital muscular dystrophy, for which there is no
Ko Tsutsui et al.
The Journal of biological chemistry, 285(7), 4870-4882 (2009-11-27)
ADAMTS (A disintegrin and metalloproteinase with thrombospondin motifs)-like (ADAMTSL) proteins, a subgroup of the ADAMTS superfamily, share several domains with ADAMTS proteinases, including thrombospondin type I repeats, a cysteine-rich domain, and an ADAMTS spacer, but lack a catalytic domain. We
Dipti Thakkar et al.
Connective tissue research, 55(5-6), 397-402 (2014-08-29)
There is increasing evidence for a progressive extracellular matrix change in rotator cuff disease progression. Directly surrounding the cell is the pericellular matrix, where assembly of matrix aggregates typically occurs making it critical in the response of tendon cells to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service