Skip to Content
Merck
All Photos(1)

Key Documents

HTS233LRTA

Sigma-Aldrich

Ready-to-Assay LH Glycoprotein Hormone Receptor Frozen Cells

Human

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.81

product name

Ready-to-Assay LH Glycoprotein Hormone Receptor Frozen Cells,

biological source

human

Quality Level

manufacturer/tradename

Ready-to-Assay

technique(s)

activity assay: suitable (cAMP)
calcium flux assay: suitable

GenBank accession no.

shipped in

dry ice

General description

EMD Millipore’s Ready-to-Assay GPCR frozen cells are designed for simple, rapid calcium assays with no requirement for intensive cell culturing. EMD Millipore has optimized the freezing conditions to provide cells with high viability and functionality post-thaw. The user simply thaws the cells and resuspends them in media, dispenses cell suspension into assay plates and, following over night recovery, assays for calcium response.

EMD Millipore’s ChemiBrite cells co-express a GPCR along with a novel variant of clytin, a calcium-activated photoprotein, to enable sensitive luminescent detection of ligand-induced calcium flux. The ChemiBrite version of clytin contains a mutation that increases its affinity for calcium to a level that permits detection of cytosolic calcium in many cells with greater sensitivity than other mitochondrially expressed photoproteins. Luminescent calcium assays offer several advantages over fluorescent calcium assays including; lower substrate cost, increased sensitivity, and lack of interference from fluorescent compounds.

Luteinizing Hormone (LH), also known as lutropin, is produced by the pituitary gland and is critical for fertility and reproduction. In women, LH modulates ovulation. In men, LH stimulates testosterone production. Several naturally occurring mutations in the LH receptor gene have been associated with human reproductive disorders (Themmen et al.). Together with TSH, hCG and FSH, LH is a member of the glycoprotein hormone family. Each member shares a common α subunit, whereas the β subunit confers functional specificity. Upon activation, LH signals primarily through the Gαqs/adenylyl cyclase/cAMP/PKA pathway as has been shown in native granulosa cells. EMD Millipore’s cloned human LH receptor-expressing ChemiBrite cells were made by recombinant transfection of HEK293 cells with ChemiBrite clytin, the LH receptor and a promiscuous G protein to couple the receptor to the calcium signaling pathway. The cells have been cryopreserved at an optimal time post-transfection. Upon thaw, recovery, and loading, the cells are ready for luminescent, fluorescent and cAMP accumulation analysis of agonists, antagonists and modulators at the LH receptor.

Cell Line Description

GPCR Cell Lines
HEK293

Biochem/physiol Actions

GPCR Class: A
Protein Target: LH
Target Sub-Family: Glycoprotein hormone

Components

Pack contains 2 vials of mycoplasma-free cells, 1 ml per vial.

Fifty (50) mL of Media Component.

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges {HCompany} to request certain information about you and the establishment where the GMOs are being handled. Click here for Enduser Declaration (EUD) Form.

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

X C Jia et al.
Molecular endocrinology (Baltimore, Md.), 5(6), 759-768 (1991-06-01)
Studies on human LH receptors are difficult due to the limited availability of clinical samples. Recent cloning of rat and porcine LH receptor cDNAs indicated that these binding sites are single polypeptides of the G-protein-coupled receptor family with seven transmembrane
Themmen APN et al.
Endocrine reviews, 21(5), 551-583 (2000-10-21)
The recent unraveling of structures of genes for the gonadotropin subunits and gonadotropin receptors has provided reproductive endocrinologists with new tools to study normal and pathological functions of the hypothalamic-pituitary-gonadal axis. Rare inactivating mutations that produce distinctive phenotypes of isolated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service