Skip to Content
Merck
All Photos(1)

Key Documents

EZHR

Millipore

Human Resistin ELISA Kit

measures and quantifies Resistin levels in 20 μL serum or plasma

Synonym(s):

Resistin ELISA Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.32

Product Name

Human Resistin ELISA, This Human Resistin ELISA is used to measure & quantify Resistin levels in Metabolism & Endocrine research.

Quality Level

species reactivity

human

packaging

kit of 1 × 96 wells

parameter

20 μL sample volume (4hr assay)

assay range

accuracy: 90-108%
linearity: 83-100%
sensitivity: 0.16 ng/mL
standard curve range: 0.16-10 ng/mL

technique(s)

ELISA: suitable

input

sample type serum
sample type plasma (K2 EDTA)

NCBI accession no.

UniProt accession no.

application(s)

research use

detection method

colorimetric (450nm/590nm)

shipped in

wet ice

storage temp.

2-8°C

General description

Resistin is a small protein (114aa) secreted from adipose tissue and circulates as a homodimer of two peptides joined by a disulfide bridge.

Specificity

Cross Reactivty
No cross reactivity with mouse resistin

Application

Research Category
Metabolism
Research Sub Category
Obesity

Metabolic Disorders
Room temperature, 3.5 hour assay 20 μL sample size
Used to detect/quantify: Resistin

Disclaimer

For research use only. Not for use in diagnostic procedures.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Skin Sens. 1 - STOT RE 2 - STOT SE 3

Target Organs

Respiratory system, Respiratory Tract

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elevated serum acylated (biologically active) ghrelin and resistin levels associate with pregnancy-induced weight gain and insulin resistance.
Eva Palik,Eva Baranyi,Zsolt Melczer,Maria Audikovszky,Albert Szocs,Gabor Winkler,Karoly Cseh
Diabetes Research and Clinical Practice null
Yen Hung Lin et al.
Clinica chimica acta; international journal of clinical chemistry, 412(9-10), 730-734 (2011-01-05)
The mechanism of obesity leading to endothelial function is complex, and involves many adipokines and inflammatory cytokines. The data is especially lacking in obese patients without metabolic syndrome. We assessed the relationship among endothelial dysfunction, anthropometric indices, adipokines and inflammatory
Serum resistin concentrations in children with Kawasaki disease.
Hiroki Nozue,Hironori Imai,Hisako Saitoh,Takeshi Aoki,Kunio Ichikawa,Tomohiro Kamoda
Inflammation Research null
Serum resistin concentrations in growth hormone-deficient children during growth hormone replacement therapy.
Hiroki Nozue,Tomohiro Kamoda,Akira Matsui
Metabolism, Clinical and Experimental null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service