Skip to Content
Merck
All Photos(1)

Key Documents

EX0245

Supelco

Ethyl acetate

HPLC, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs

Synonym(s):

Acetic Acid Ethyl Ester, EtOAc

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOC2H5
CAS Number:
Molecular Weight:
88.11
Beilstein:
506104
MDL number:
UNSPSC Code:
12352108
EC Index Number:
205-500-4
grade:
reagent
technique(s):
HPLC: suitable
bp:
76.5-77.5 °C (lit.)
vapor pressure:
73 mmHg ( 20 °C)

grade

reagent

Quality Level

Agency

meets USP/NF

vapor density

3 (20 °C, vs air)

vapor pressure

73 mmHg ( 20 °C)

Assay

≥99.8% (GC)

form

liquid

autoignition temp.

801 °F

expl. lim.

2.2-11.5 %, 38 °F

technique(s)

HPLC: suitable

impurities

≤0.0009 meq/g Titrable acid
≤0.05% Water (H2O)

evapn. residue

≤5 ppm

color

APHA: ≤10

refractive index

n20/D 1.3720 (lit.)

bp

76.5-77.5 °C (lit.)

mp

−84 °C (lit.)

transition temp

flash point -4 °C

density

0.902 g/mL at 25 °C (lit.)

UV cutoff

≤255 nm

UV absorption

λ: 255 nm Amax: ≤1.00
λ: 257 nm Amax: ≤0.50
λ: 263 nm Amax: ≤0.10
λ: 275 nm Amax: ≤0.05
λ: 300-400 nm Amax: ≤0.01

format

neat

shipped in

ambient

storage temp.

room temp

SMILES string

CCOC(C)=O

InChI

1S/C4H8O2/c1-3-6-4(2)5/h3H2,1-2H3

InChI key

XEKOWRVHYACXOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

EX0245P-1 is packaged in poly coated glass bottles
EX0245-NP10 is sold as 2 x 10 L
HPLC, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs

Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

AppearanceClear liquid, free from particulatesAssay (GC)99.8% minColor (APHA)10 maxEvaporation residue5ppm maxFiltered through 0.2 µm filterTo pass testFormLiquidInfrared SpectrumConforms to standardSubstances darkened by sulfuric acidTo pass testTitrable acid0.0009meq/g maxUV Abs. at 255 nm1.00AU maxUV Abs. at 257 nm0.50AU maxUV Abs. at 263 nm0.10AU maxUV Abs. at 275 nm0.05AU maxUV Abs. at 300 - 4000.01AU maxUV Cut-off255nm maxWater (H2O)0.05% max

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

24.8 °F - closed cup

Flash Point(C)

-4 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service