Skip to Content
Merck
All Photos(1)

Key Documents

CX1723

Supelco

Citric Acid, Anhydrous

anhydrous Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Synonym(s):

Citric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC(COOH)(CH2COOH)2
CAS Number:
Molecular Weight:
192.12
Beilstein:
782061
MDL number:
UNSPSC Code:
12352106
EC Index Number:
201-069-1

Agency

meets USP/NF

Quality Level

vapor pressure

<0.1 hPa ( 20 °C)

Assay

99.5-100.5% dry basis (alkalimetric)

form

fine powder

expl. lim.

8 %, 65 °F

impurities

≤0.005% Water-insoluble matter
≤1% Water (Karl Fischer)

sulfated ash (600 °C)

≤0.02%

color

white to almost white

pH

1.7 (20 °C, 100 g/L in H2O)

pKa 

(1) 3.13, (2) 4.76, (3) 6.4

bp

200 °C/1013 hPa (decomposition)

mp

153-159 °C (lit.)

solubility

1330 g/L

density

1.67 g/cm3 at 20 °C

anion traces

chloride (Cl-): ≤0.001%
oxalate (C2O42-): ≤0.036%
phosphate (PO43-): ≤0.001%
sulfate (SO42-): ≤0.002%

cation traces

Fe: ≤0.0003%
Pb: ≤0.0002%

shipped in

ambient

storage temp.

room temp

SMILES string

OC(=O)CC(O)(CC(O)=O)C(O)=O

InChI

1S/C6H8O7/c7-3(8)1-6(13,5(11)12)2-4(9)10/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)

InChI key

KRKNYBCHXYNGOX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

anhydrous Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Analysis Note

Appearancewhite to almost white, fine powderAssay (alkalimetric on dried substance)99.5-100.5%Chloride (Cl)0.001% maxIdentity (IR-spectrum)passes testIron (Fe)0.0003% maxLead (Pb)0.0002% maxOxalate (C2O4)0.036% maxPhosphate (PO4)0.001% maxReadily carbonizable substancespasses testSulfate (SO4)0.002% maxSulfated ash (600°C)0.02% maxWater (acc. to Karl Fischer)1.0% maxWater-insoluble matter0.005% max

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service