Skip to Content
Merck
All Photos(1)

Key Documents

CX0120

Supelco

Calcium carbonate

Synonym(s):

Carbonic Acid Calcium Salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaCO3
CAS Number:
Molecular Weight:
100.09
Beilstein:
8008338
MDL number:
UNSPSC Code:
12352302
EC Index Number:
207-439-9
grade:
for analytical purposes
form:
powder
solubility:
0.017 g/L

grade

for analytical purposes

Quality Level

Assay

98-100.5% dry basis

form

powder

impurities

≤0.2% Acid-insoluble substances

loss

≤2% loss on drying

color

white

pH

9.5-10.5 (20 °C, 100 g/L in H2O, slurry)

mp

825 °C (decomposition)

solubility

0.017 g/L

density

2.93 g/mL at 25 °C (lit.)

cation traces

Ba:, passes test
heavy metals (as Pb): ≤0.003%
magnesium and alkali salts (as sulfates): ≤1.0%

shipped in

ambient

storage temp.

room temp

SMILES string

[Ca++].[O-]C([O-])=O

InChI

1S/CH2O3.Ca/c2-1(3)4;/h(H2,2,3,4);/q;+2/p-2

InChI key

VTYYLEPIZMXCLO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Acid-insoluble substances: 0.2% max
Assay (dried): 98-100.5%
Barium (Ba): To pass test
Color: White
Description: To pass test
Form: Powder
Heavy metals (as Pb): 0.003% max
Identification: To pass test
Loss on drying: 2.0% max
Magnesium and alkali salts: 1.0% max
Solubility: To pass test

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service