Skip to Content
Merck
All Photos(1)

Documents

AP307P

Sigma-Aldrich

Goat Anti-Rabbit IgG Antibody, (H+L) HRP conjugate

1 mg/mL, Chemicon®

Synonym(s):

Anti-Rabbit IgG HRP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

peroxidase conjugate

antibody form

affinity purified immunoglobulin

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

rabbit

manufacturer/tradename

Chemicon®

concentration

1 mg/mL

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

shipped in

wet ice

target post-translational modification

unmodified

Related Categories

General description

Immunoglobulin G (IgG) belongs to the immunoglobulin family and is a widely expressed serum antibody. An immunoglobulin has two heavy chains and two light chains connected by a disulfide bond. It is a glycoprotein and mainly helps in immune defense. IgG is a major class of immunoglobulin. Rabbit IgG is further divided into five classes- IgM, IgG, IgA, IgD and IgE. It has only one IgG variant with one interheavy disulfide bridge.

Specificity

Specific for rabbit IgG, heavy and light chain. The cross-reactivities of anti-rabbit IgG antibody are tested in an ELISA. Minimum cross-reactivity to human IgG.

Application

Goat Anti-Rabbit IgG Antibody, (H+L) HRP conjugate has been used in:
  • immunohistochemistry (IHC)
  • western blotting
  • immunoprecipitation

Biochem/physiol Actions

Immunoglobulin G (IgG) participates in hypersensitivity type II and type III. It helps in opsonization, complement fixation and antibody dependent cell mediated cytotoxicity.

Quality

Western Blot, chromogenic:
1:200-1:500 dilution can be used (Coligan, J., 1997; Harlow, E., 1988).

Western Blot, chemiluminescent:
1:2,000-1:5,000 dilution can be used (Coligan, J., 1997).

Physical form

ImmunoAffinity Purified
Purified Goat IgG conjugated to horseradish peroxidase in buffer containing liquid in 0.01M PBS, pH 7.1 with 15mg/mL BSA and 0.1% Kathon.

Storage and Stability

The undiluted antibody is stable at 2-8°C for 12 months. Do not store in a diluted format.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stabilization of microtubular cytoskeleton protects neurons from toxicity of N-terminal fragment of cytosolic prion protein
Zajkowski T, et al.
Biochimica et Biophysica Acta, 1853(10), 2228-2239 (2015)
Transsynaptic activity-dependent regulation of axon branching and neurotrophin expression in vivo.
Calinescu, AA; Liu, T; Wang, MM; Borjigin, J
The Journal of Neuroscience null
Phospho-Delta-Np63alpha-responsive microRNAs contribute to the regulation of necroptosis in squamous cell carcinoma upon cisplatin exposure
Ratovitski EA
Febs Letters, 589(12), 1352-1358 (2015)
Bisdemethoxycurcumin attenuates gastric adenocarcinoma growth by inducing mitochondrial dysfunction
Luo C, et al.
Oncology Letters, 270-274 (2015)
Tomas Blanco-Mezquita et al.
Investigative ophthalmology & visual science, 54(6), 3880-3890 (2013-05-04)
Nerve growth factor (NGF) is a neuropeptide essential for the development, survival, growth, and differentiation of corneal cells. Its effects are mediated by both TrkA and p75 receptors. Clinically relevant use of NGF was introduced to treat neurotrophic ulcerations in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service