Skip to Content
Merck
All Photos(2)

Key Documents

ABC438

Sigma-Aldrich

Anti-PKM2 Antibody

serum, from rabbit

Synonym(s):

Pyruvate kinase PKM, Cytosolic thyroid hormone-binding protein, CTHBP, Opa-interacting protein 3, OIP-3, Pyruvate kinase 2/3, Pyruvate kinase muscle isozyme, Thyroid hormone-binding protein 1, THBP1, Tumor M2-PK, p58

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, rat, mouse

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PKM(5315)

General description

Pyruvate kinase isozymes M1/M2 (PKM2) is also called Cytosolic thyroid hormone-binding protein (CTHBP), Opa-interacting protein 3 (OIP-3), Pyruvate kinase 2/3, Pyruvate kinase muscle isozyme, Thyroid hormone-binding protein 1 (THBP1) and p58. PKM2 is a glycolytic enzyme that assists in generating ATP. PKM2 also stimulates POU5F1-mediated transcriptional activation, and plays a role in caspase independent cell death of tumor cells. The ratio between the highly active tetrameric form and nearly inactive dimeric form determines whether glucose carbons are channeled to biosynthetic processes or used for glycolytic ATP production. This control of glycolysis and is important for tumor cell proliferation and survival. PKM2 is specifically expressed in proliferating cells, such as embryonic stem cells, embryonic carcinoma cells, and cancer cells.

Specificity

This antibody is specific to PKM2.

Immunogen

Linear peptide corresponding to human PKM2.

Application

Anti-PKM2 Antibody detects level of PKM2 & has been published & validated for use in WB and IHC.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected PKM2 in 10 µg of C2C12, HEK293, HeLa, HepG2, human placenta, HUVEC, Jurkat, L6, mouse brain, NIH/3T3, PC12, PC3, and RAW264.7 lysate.
Western Blotting Analysis: A representative lot detected PKM2 in HEK293 cell lysate (Christofk, R. H., et al. (2008). Nature. 452:230-233).
Immunohistochemistry Analysis: A representative lot detected PKM2 in HEK293 tissue (Christofk, R. H., et al. (2008). Nature. 452:230-233).

Quality

Evaluated by Western Blotting in A431 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected PKM2 in 10 µg of A431 cell lysate.

Target description

~ 58 kDa observed

Physical form

Rabbit polyclonal serum with 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service