Skip to Content
Merck
All Photos(2)

Key Documents

AB5060

Sigma-Aldrich

Anti-Substance P Receptor Antibody, pain

serum, Chemicon®

Synonym(s):

NK-1 Receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, guinea pig, mouse

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... TAC1(6863)

General description

Substance P is a tachykinin family neuropeptide that functions as a neurotransmitter and neuromodulator. In the CNS, Substance P is involved in the regulation of mood, anxiety, sleep, saiety, reinforcement, respiratory rhythm and pain processing. In nociception, the neuropeptide is released at local spinal cord synapses. Recent research now shows substance P is potentially involved in endothelial cell differentiation as well as diverse roles in neuroinflammatory response. The endogenous receptor for Substance P is neurokinin 1 receptor (NK1 receptor), which belongs to the tachykinin receptor sub-family of GPCRs.

Specificity

Specific for the Substance P Receptor (NK-1).

Immunogen

Epitope: C terminus
Synthetic peptide that corresponds to a 23 amino acid sequence (385-407) of the COOH terminus of the rat Substance P Receptor (NK-1).

Application

Anti-Substance P Receptor Antibody, pain detects level of Substance P Receptor & has been published & validated for use in IF, IH & WB.
Immunohistochemistry:
A previous lot was used at 1:100-1:1,000 dilution.

Western Blot Analysis:
A previous lot was used at 1:500 dilution.

Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Neuroinflammation & Pain

Quality

Evaluated by western blot on Mouse brain membrane lysates.

Western Blot Analysis:
1:500 dilution of this antibody detected NK-1R on 10 μg of Mouse brain membrane lysates.

Target description

35 kDa

Linkage

Replaces: AB5800

Physical form

Rabbit polyclonal serum containing no preservative.
Unpurified

Storage and Stability

Stable for 1 year at -20ºC from date of receipt.

Analysis Note

Control
Spinal Cord, brain membrane lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Terminal field and firing selectivity of cholecystokinin-expressing interneurons in the hippocampal CA3 area.
Lasztoczi, B; Tukker, JJ; Somogyi, P; Klausberger, T
The Journal of Neuroscience null
Yanyan Zhang et al.
International journal of oral science, 11(3), 24-24 (2019-09-11)
Inflammatory orofacial pain, in which substance P (SP) plays an important role, is closely related to the cross-talk between trigeminal ganglion (TG) neurons and satellite glial cells (SGCs). SGC activation is emerging as the key mechanism underlying inflammatory pain through
L Sosulina et al.
Journal of neurophysiology, 114(4), 2500-2508 (2015-09-04)
Substance P (SP) is implicated in stress regulation and affective and anxiety-related behavior. Particularly high expression has been found in the main output region of the amygdala complex, the central amygdala (CE). Here we investigated the cellular mechanisms of SP
Amie Severino et al.
Pain, 159(8), 1607-1620 (2018-04-21)
The latent sensitization model of chronic pain reveals that recovery from some types of long-term hyperalgesia is an altered state in which nociceptive sensitization persists but is suppressed by the ongoing activity of analgesic receptors such as μ-opioid receptors (MORs).
Marco Bocchio et al.
eNeuro, 3(5) (2016-11-09)
Unraveling the roles of distinct neuron types is a fundamental challenge to understanding brain function in health and disease. In the amygdala, a brain structure regulating emotional behavior, the diversity of GABAergic neurons has been only partially explored. We report

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service