Skip to Content
Merck
All Photos(2)

Key Documents

AB1858

Sigma-Aldrich

Anti-Osteonectin Antibody

serum, Chemicon®

Synonym(s):

SPARC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable (paraffin)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SPARC(6678)

General description

Osteonectin is one of the major noncollagenous proteins in the extracellular matrix of bone. It is a glycoprotein with a molecular weight of 43 kDa and has calcium-binding properties.

Specificity

Human Osteonectin (ON).

Immunogen

recombinant osteonectin expressed in human embryonal kidney line HEK-293.

Application

Anti-Osteonectin Antibody is an antibody against Osteonectin for use in ELISA, IH(P).
ELISA (1:5,000)

Immunohistochemistry on paraffin-embedded tissue sections (1:1,000)

Optimal working dilutions must be determined by the end user.
Research Category
Cell Structure
Research Sub Category
Osteobiology

Physical form

Rabbit serum. Lyophilized. Reconstitute with 100 μL of sterile distillled water.

Storage and Stability

Maintain lyophilized material at -20°C for up to 12 months. After reconstitution maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eriberto Bressan et al.
International journal of molecular and cellular medicine, 4(4), 197-208 (2016-03-26)
The biological interaction between the jaw bones and dental implant is fundamental for the long-term success of dental implant placement. Nevertheless, the insufficient bone volume remains a major clinical problem, especially in case of immediate dental implant. Using a canine
Shane A Liddelow et al.
PloS one, 7(3), e33554-e33554 (2012-03-30)
Exchange mechanisms across the blood-cerebrospinal fluid (CSF) barrier in the choroid plexuses within the cerebral ventricles control access of molecules to the central nervous system, especially in early development when the brain is poorly vascularised. However, little is known about
Letizia Ferroni et al.
Journal of tissue engineering and regenerative medicine, 11(5), 1332-1342 (2015-06-09)
This study investigated the effect of extremely low-frequency pulsed electromagnetic fields (PEMFs) on skin wound healing in an in vitro dermal-like tissue. In this study, fibroblast and endothelial cells were utilized for the in vitro reconstruction of dermal-like tissues treated
Shane A Liddelow et al.
PloS one, 9(9), e106592-e106592 (2014-09-12)
To maintain the precise internal milieu of the mammalian central nervous system, well-controlled transfer of molecules from periphery into brain is required. Recently the soluble and cell-surface albumin-binding glycoprotein SPARC (secreted protein acidic and rich in cysteine) has been implicated
Luigi Clauser et al.
PloS one, 9(11), e110796-e110796 (2014-11-07)
Structural fat grafting utilizes the centrifugation of liposuction aspirates to create a graded density of adipose tissue. This study was performed to qualitatively investigate the effects of centrifugation on stem cells present in adipose tissue. Liposuction aspirates were obtained from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service