Skip to Content
Merck
All Photos(1)

Key Documents

8.43861

Sigma-Aldrich

Lauroyl peroxide

for synthesis

Synonym(s):

Lauroyl peroxide

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C24H46O4
CAS Number:
Molecular Weight:
398.62
MDL number:
UNSPSC Code:
12352120
EC Index Number:
203-326-3
NACRES:
NA.22

Quality Level

form

powder

autoignition temp.

112 °C

potency

10000 mg/kg LD50, oral (Rat)

mp

49 °C

density

0.91 g/cm3 at 20 °C

storage temp.

15-25°C

InChI

1S/C24H46O4/c1-3-5-7-9-11-13-15-17-19-21-23(25)27-28-24(26)22-20-18-16-14-12-10-8-6-4-2/h3-22H2,1-2H3

InChI key

YIVJZNGAASQVEM-UHFFFAOYSA-N

Analysis Note

Assay (iodometric): ≥ 98.0 %
Identity (IR): passes test

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Org. Perox. D

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service