Skip to Content
Merck
All Photos(1)

Key Documents

8.22069

Sigma-Aldrich

Methylhydroxyethylcellulose

for synthesis

Synonym(s):

Methylhydroxyethylcellulose

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

form

powder

autoignition temp.

>360 °C

potency

>2000 mg/kg LD50, oral (Rat)

density

1.28-1.30 g/cm3 at 20 °C

bulk density

350 kg/m3

storage temp.

2-30°C

SMILES string

O3[C@H]([C@@H]([C@H]([C@@H]([C@H]3COC)OC)OC)OC)O[C@@H]4[C@H](O[C@H]([C@@H]([C@H]4OC)OC)OC)COC.O1[C@H]([C@@H]([C@H]([C@@H]([C@H]1CO)O)O)O)O[C@@H]2[C@H](O[C@H]([C@@H]([C@H]2O)O)O)CO.OCCO

InChI key

JTWZVKMWOCBJEV-RXUSYSPUSA-N

Analysis Note

Activeassay: ≥ 92.0 %
Loss on drying: lot specific result
Assay (NaCl): lot specific result
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service