Skip to Content
Merck
All Photos(1)

Key Documents

8.22057

Sigma-Aldrich

Gluconic acid

(50% solution in water) for synthesis

Synonym(s):

2,3,4,5,6-Pentahydroxycaproic acid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352106
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

form

liquid

pH

2.2 (20 °C, 500 g/L in H2O)

bp

105-106 °C/1013 hPa

density

1.24 g/cm3 at 20 °C

storage temp.

2-30°C

Related Categories

Application

  • Biodegradable chelating agent for wastewater treatment: Gluconic acid serves as a biodegradable chelating agent that enhances metal ion removal in wastewater treatment processes. This application is valuable for environmental sustainability and industrial wastewater management (Galaz et al., 2024).
  • Pharmaceutical production optimization: Gluconic acid is used in pharmaceutical production, where its properties help in the optimization of medication formulations. This application ensures effective drug delivery and stability, crucial for therapeutic efficacy (Xing et al., 2024).
  • Food preservation: In the food industry, gluconic acid is applied as a natural preservative, enhancing the shelf life and safety of food products. Its use supports food safety regulations and consumer health (Lu et al., 2024).
  • Enhancement of bioenergy production: Gluconic acid contributes to the enhancement of bioenergy production processes, particularly in biofuel production through fermentation optimization techniques. This application is key to advancing renewable energy technologies (Liu et al., 2024).
  • Research tool in metabolomics: Utilized as a research tool in metabolomics, gluconic acid aids in the study of metabolic processes in various biological systems, providing insights into cellular functions and disease mechanisms (Patakova et al., 2024).

Analysis Note

Assay (acidimetric): 48.0 - 52.0 %
Density (d 20 °C/ 4 °C): 1.230 - 1.245

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. What is the grade of this product?

    1 answer
    1. This product is intended for research use only and is assigned a quality level of 200. Please see the link below to review additional information regarding the M-Clarity guide to product quality:\
      https://www.sigmaaldrich.com/life-science/quality-and-regulatory-management/m-clarity-program?srsltid=AfmBOoq7n755YAOZopvcX0WoIMiGDwZ3npVU_1zNdtN7NAFm6iNNvpcf

      Helpful?

  2. Is the glucon acid in excipient quality?

    1 answer
    1. Product No. 8.22057 is not considered a pharma grade excipient. It is intended for synthesis in research applications.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service