Skip to Content
Merck
All Photos(1)

Key Documents

8.18896

Sigma-Aldrich

1-Bromotetradecane

for synthesis

Synonym(s):

1-Bromotetradecane, Myristyl bromide, Tetradecyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)13Br
CAS Number:
Molecular Weight:
277.28
MDL number:
UNSPSC Code:
12352101
EC Index Number:
203-999-3

Assay

≥97.0% (GC)

form

liquid

bp

175-178 °C/27 hPa

mp

4-6 °C

transition temp

flash point 149 °C

density

1.02 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C14H29Br/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15/h2-14H2,1H3

InChI key

KOFZTCSTGIWCQG-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.015 - 1.020
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(C)

113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service