Skip to Content
Merck
All Photos(1)

Key Documents

8.18311

Sigma-Aldrich

Copper(I) iodide

for synthesis

Synonym(s):

Copper(I) iodide, Copper monoidide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CuI
CAS Number:
Molecular Weight:
190.45
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-674-6
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

form

solid

bp

1290 °C/1013 hPa

mp

588 °C

solubility

0.08 g/L

density

5.62 g/cm3 at 20 °C

bulk density

1400 kg/m3

storage temp.

2-30°C

SMILES string

[Cu]I

InChI

1S/Cu.HI/h;1H/q+1;/p-1

InChI key

LSXDOTMGLUJQCM-UHFFFAOYSA-M

Analysis Note

Assay (iodometric): ≥ 98.0 %

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1A - STOT RE 1 Oral

Target Organs

Thyroid

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service