Skip to Content
Merck
All Photos(1)

Key Documents

8.15100

Sigma-Aldrich

2,5-Diethyl-1,3,4-thiadiazole

for synthesis

Synonym(s):

2,5-Diethyl-1,3,4-thiadiazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10N2S
CAS Number:
Molecular Weight:
142.22
MDL number:
UNSPSC Code:
12352005
EC Index Number:
255-143-3
NACRES:
NA.22

Quality Level

Assay

≥96.0% (GC)

form

liquid

bp

105 °C/18.7 hPa

density

1.071 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[s]1c(nnc1CC)CC

InChI

1S/C6H10N2S/c1-3-5-7-8-6(4-2)9-5/h3-4H2,1-2H3

InChI key

BXVZVOBTUSLCIO-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 96.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.068 - 1.073
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service