Skip to Content
Merck
All Photos(1)

Key Documents

8.04726

Sigma-Aldrich

Indole-3-carbaldehyde

for synthesis

Synonym(s):

Indole-3-carbaldehyde, Indol-3-aldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7NO
CAS Number:
Molecular Weight:
145.16
MDL number:
UNSPSC Code:
12352114
EC Index Number:
207-665-8
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥98.0% (acidimetric)

autoignition temp.

>550 °C

mp

195-198 °C

transition temp

flash point 240 °C

bulk density

200‑300 kg/m3

storage temp.

2-30°C

SMILES string

[nH]1c2c(c(c1)C=O)cccc2

InChI

1S/C9H7NO/c11-6-7-5-10-9-4-2-1-3-8(7)9/h1-6,10H

InChI key

OLNJUISKUQQNIM-UHFFFAOYSA-N

Analysis Note

Assay (acidimetric): ≥ 98.0 %
Melting range (lower value): ≥ 193 °C
Melting range (upper value): ≤ 197 °C
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service