Skip to Content
Merck
All Photos(2)

Key Documents

662210

Sigma-Aldrich

USP14 Inhibitor, IU1

The USP14 Inhibitor, IU1 controls the biological activity of USP14. This small molecule/inhibitor is primarily used for Cell Structure applications.

Synonym(s):

USP14 Inhibitor, IU1, 1-(1-(4-Fluorophenyl)-2,5-dimethyl-1H-pyrrol-3-yl)-2-(pyrrolidin-1-yl)ethanone, 1-(1-(4-Fluorophenyl)-2,5-dimethylpyrrol-3-yl)-2-pyrrolidin-1-ylethanone, Ubiquitin Specific Protease 14 Inhibitor, DUB Inhibitor IX

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H21FN2O
Molecular Weight:
300.37
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥90% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

beige

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

Fc1ccc(cc1)[n]2c(c(cc2C)C(=O)CN3CCCC3)C

InChI

1S/C18H21FN2O/c1-13-11-17(18(22)12-20-9-3-4-10-20)14(2)21(13)16-7-5-15(19)6-8-16/h5-8,11H,3-4,9-10,12H2,1-2H3

InChI key

JUWDSDKJBMFLHE-UHFFFAOYSA-N

General description

A cell-permeable pyrrolylpyrrolidinyl-ethanone that inhibits the proteasome association-dependent USP14 deubiquitinase (DUB) activity in a reversible manner (IC50 = 4.7 µM against Ub-AMC hydrolysis), exhibiting little or no activity toward 8 other DUBs. Shown to downregulate tau, TDP-43, ATXN3, GFAP, and free Ub levels in MEF cultures in a dose- (25 to 100 µM), time-, and USP14-dependent manner. USP14-catalyzed trimming of protein surface polyubiquitin chains rescues proteasome-bound proteins from proteolytic degradation and thereby prevents the depletion of free ubiquitin pool in cells. Comparing to b-AP15 (Cat. No. 662140), IU1 does not inhibit UCH37/UCH-L5 (IC50 = 700 µM).

Packaging

Packaged under inert gas

Warning

Toxicity: Regulatory Review (Z)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Lee, B.H., et al. 2010. Nature467, 179.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service