Skip to Content
Merck
All Photos(2)

Key Documents

5C44

Millipore

NovaSeptum® GO Conical Tube Sampling Unit, Manifold (5-valve) with Inlet C-Flex® Tubing

sterile; γ-irradiated, volume × volume 15 mL × 50 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104019

material

TPE tubing (Inlet C-Flex)
polycarbonate Luer lock
polypropylene clip (TTP clip)
polypropylene tube (Fluid contact layer (conical tube))
polystyrene tube (Fluid contact layer (conical tube))
tubing (Female and male Luer-Lok®)

Quality Level

Agency

according to ISO 11137 (sterilization)
according to ISO Clean Room Class 8 conditions in a facility
certified by the ISO 14644-1 standards
meets USP 88 biological reactivity (all component materials)

sterility

sterile; γ-irradiated

product line

NovaSeptum® GO

feature

autoclavable: no

parameter

max. pressure at 0.50 bar (7.25 psi)
2-60 °C temp. range (36-140 °F)

volume × volume

15 mL × 50 mL

application(s)

cell therapy
gene therapy
life science and biopharma
mAb
ophthalmics
parenterals
pharma/biopharma processes
sterile sampling
viral therapy

storage temp.

2-30°C

General description

Device configuration: single Unit Sampling
NovaSeptum® GO Conical Tube Sampling Unit, Manifold (5-valve) with Inlet C-Flex® Tubing are small volume sampling containers that fit our established NovaSeptum® GO sampling system. These sampling containers are available in two sizes: 15 mL polystyrene tubes ideal for endotoxin testing, and 50 mL polypropylene tubes for bioburden and other physico-chemical tests. These conical tubes provide flexibility to manufacturers for sampling at any point in the production process eliminating the need to aliquot.

When used in a single-use manufacturing process, a weldable option with C-Flex® tubing is available.

The NovaSeptum® GO Conical Tube Sampling Unit offers you flexibility while minimizing the risk of both product and process contamination.

Application

Sterility testing, pyrogenic testing, endotoxin testing, chemical analysis, pH analysis, fermentation applications or any other routine lab testing
supported liquid extraction

Features and Benefits

Fluid contact layer made of polystyrene, ideal for endotoxin testingSmall size, no need to aliquot, Manifold Format, C-Flex inlet weldable

Packaging

8 units per box

Components

Luer: Polycarbonate

Fluid contact layer (conical tube): Polystyrene (15mL) and Polypropylene (50mL), dip tube: LDPE (Low density polyethylene pharmalene FL20PH)

TTP clip Polypropylène (15mL only)

Tubing: Thermoplastic elastomer (TPE), Inlet C-Flex

Outlet Tubing: Female and male Luer-Lok®

Legal Information

C-Flex is a registered trademark of Saint-Gobain Performance Plastics Corp.
Luer-Lok is a registered trademark of Becton-Dickinson & Co.
NOVASEPTUM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service