Skip to Content
Merck
All Photos(1)

Key Documents

567022

Sigma-Aldrich

Simvastatin sodium salt

≥95% (HPLC), liquid, L-type calcium channel blocker, Calbiochem®

Synonym(s):

InSolution Simvastatin, Sodium Salt, L-Type Calcium Channel Blocker VI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H39O6 · Na
Molecular Weight:
458.56
UNSPSC Code:
51111800
NACRES:
NA.77

Product Name

Simvastatin, Sodium Salt, InSolution, ≥95%

Quality Level

Assay

≥95% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
protect from light

shipped in

ambient

storage temp.

−70°C

General description

Carboxylate form of Simvastatin (Cat. No. 567020) that is active in whole cells and in cell-free assays. A lipophilic 3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) reductase inhibitor that blocks Ras function through inhibition of farnesylation. Also inhibits glucose-induced Ca2+-signaling and insulin secretion by blocking L-type Ca2+ channels in rat islet β-cells. Inhibits cell proliferation of human smooth muscle cells. The solid form of this compound (Cat. No. 567021) is also available.

Biochem/physiol Actions

Cell permeable: yes
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Physical form

A 10 mM (5 mg/1.09 ml) solution of Simvastatin, Sodium Salt (Cat. No. 567021) in H₂O.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Yada, T., et al. 1999. Br. J. Pharmacol.126, 1205.
Clutterbuck, R.D., et al. 1998. Br. J. Haematol.102, 522.
Yokoyama, K., et al. 1998. Mol. Biochem. Parasitol.94, 87.
Negre-Aminou, P., et al. 1997. Biochim. Biophys. Acta1345, 259.
Vrtovsnik, F., et al. 1997. Kidney Int.52, 1016.
Keyomarsi, K., et al. 1991. Cancer Res.51, 3602.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sampath Katakam et al.
Life science alliance, 5(3) (2022-01-06)
Malignant tumors commonly display necrosis, which invariably triggers an inflammatory response that supports tumor growth. However, the effect on tumor cells of necrotic debris, or damage-associated molecular patterns (DAMPs) released by dying cells is unknown. Here, we addressed the effect
Luipa Khandker et al.
Cell reports, 38(9), 110423-110423 (2022-03-03)
Brain and spinal cord oligodendroglia have distinct functional characteristics, and cell-autonomous loss of individual genes can result in different regional phenotypes. However, a molecular basis for these distinctions is unknown. Using single-cell analysis of oligodendroglia during developmental myelination, we demonstrate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service