Skip to Content
Merck
All Photos(1)

Key Documents

492042

Sigma-Aldrich

Nrf2 Activator III, TAT-14 Peptide

The Nrf2 Activator III, TAT-14 Peptide modulates the biological activity of Nrf2. This small molecule/inhibitor is primarily used for Biochemicals applications.

Synonym(s):

Nrf2 Activator III, TAT-14 Peptide, H-Tyr-Gly-Arg-Lys-Lys-Arg-Arg-Gln-Arg-Arg-Arg~Leu-Gln-Leu-Asp-Glu-Glu-Thr-Gly-Glu-Phe-Leu-Pro-Ile-Gln-OH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C137H230N48O39
CAS Number:
Molecular Weight:
3173.59
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.51
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A 14-mer peptide (LQLDEETGEFLPIQ) conjugated to cell-penetrating trans-activating transcriptional activator (TAT) sequence (YGRKKRRQRRRLQ) that targets the nuclear factor (erythroid-derived 2)-like 2 (Nrf2) binding site on Kelch-like ECH-associated protein 1 (Keap1). Competitively disrupts Nrf2-Keap1 interaction and stabilizes cytosolic Nrf2, and promotes its nuclear translocation and binding to the antioxidant response element (ARE). Exhibits anti-inflammatory properties and increases heme-oxygenase-1 gene expression (~75 µM). Also shown to suppress LPS-induced TNF-α secretion in THP-1 monocytes (~ 37.5 µM).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

H-Tyr-Gly-Arg-Lys-Lys-Arg-Arg-Gln-Arg-Arg-Arg~Leu-Gln-Leu-Asp-Glu-Glu-Thr-Gly-Glu-Phe-Leu-Pro-Ile-Gln-OH

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Steel, R., et al. 2012. ACS Med Chem Lett.3, 407.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service