Skip to Content
Merck
All Photos(1)

Key Documents

442611-M

Millipore

Magnesium Chloride, Hexahydrate, Molecular Biology Grade

Synonym(s):

Magnesium chloride hexahydrate, MgCl₂

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgCl2 · 6H2O
CAS Number:
Molecular Weight:
203.30
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.25
Assay:
≥99% (complexometric)
grade:
Molecular Biology
form:
crystalline solid
solubility:
water: 20 mg/mL
Pricing and availability is not currently available.

grade

Molecular Biology

Quality Level

Assay

≥99% (complexometric)

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

solubility

water: 20 mg/mL

cation traces

heavy metals (as Pb): ≤0.0005%

foreign activity

DNases, proteases, RNases, none detected

storage temp.

10-30°C

SMILES string

O.O.O.O.O.O.Cl[Mg]Cl

InChI

1S/2ClH.Mg.6H2O/h2*1H;;6*1H2/q;;+2;;;;;;/p-2

InChI key

DHRRIBDTHFBPNG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Has an optimal pH of 5.0-6.5.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, store at room remperature (20°C). Stock solutions are stable for up to 6 months at room temperature.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service