Skip to Content
Merck
All Photos(1)

Key Documents

341291

Sigma-Aldrich

Kp7-6

≥98% (HPLC), lyophilized powder, Fas/FasL antagonist, Calbiochem®

Synonym(s):

Fas/FasL Antagonist, Kp7-6, H-YC*DEHFC*Y-OH, Cyclic [Cys-Cys disulfide], Kp7-6

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H56N10O15S2
Molecular Weight:
1077.15
UNSPSC Code:
12352200
NACRES:
NA.77

product name

Fas/FasL Antagonist, Kp7-6, The Fas/FasL Antagonist, Kp7-6 controls the biological activity of Fas/FasL. This small molecule/inhibitor is primarily used for Cancer applications.

Quality Level

Assay

≥98% (HPLC)

form

lyophilized powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

An exocyclic cysteine-knot peptide that specifically antagonizes Fas/FasL-mediated cellular apoptotic signals (58% reduction of FasL-induced apoptosis in Jurkat cells at 1 mg/ml). Binds to FasL (Cat. Nos. PF033 and PF092) and Fas (CD95/APO-1) with comparable affinity (Kd = 11.2 & 13.2 µM, respectively), resulting in disabled receptor ensembles and altered signaling. Shown to prevent Con A (Cat. No. 234567)-induced liver injury in a murine hepatitis model in vivo. Does not bind to TNF-α or TNFR.
An exocyclic cystine-knot peptide that specifically antagonizes Fas/FasL-mediated cellular apoptotic signals (58% reduction of FasL-induced apoptosis in Jurkat cells at 1 mg/ml). Binds to FasL (Cat. Nos. PF033 and PF092) and Fas (CD95/APO-1) with comparable affinity (Kd = 11.2 & 13.2 µM, respectively), resulting in disabled receptor ensembles and altered signaling pathways. Shown to prevent Con A (Cat. No. 234567)-induced liver injury in a murine hepatitis model in vivo. Does not bind to TNF-α or TNFR.

Biochem/physiol Actions

Cell permeable: no
Kd = 11.2 & 13.2 µM for FasL and Fas (CD95/APO-1), respectively
Primary Target
Fas/FasL-mediated cellular apoptotic signals
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

H₂N-Tyr-Cys*-Asp-Glu-His-Phe-Cys*-Tyr-CO₂H, Cyclic [disulfide bond between Cys-Cys]

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Carmona, E.M., et al. 2006. J. Immunol.177, 459.
Hasegawa, A., et al. 2004. Proc. Natl. Acad. Sci. USA101, 6599.

Legal Information

Sold under license of U.S. Patent 7,288,519.
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thais F C Fraga-Silva et al.
iScience, 26(12), 108366-108366 (2023-12-04)
Airway epithelial cells (AEC) infected with SARS-CoV-2 may drive the dysfunction of macrophages during COVID-19. We hypothesized that the direct interaction of AEC with macrophages mediated by CD95/CD95L or indirect interaction mediated by IL-6 signaling are key steps for the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service