Skip to Content
Merck
All Photos(1)

Key Documents

322133

Sigma-Aldrich

DSP, Water-Soluble

Synonym(s):

3,3-Dithio- bis-(sulfosuccinimidyl)propionate, DTSSP, Sulfo-DSP, 3,3-Dithio-bis-(sulfosuccinimidyl)propionate, DTSSP, Sulfo-DSP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116133
NACRES:
NA.24
Pricing and availability is not currently available.

Assay

≥94% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

off-white to light tan

solubility

water: 6 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

Bifunctional cross-linker. The disulfide bond can be cleaved with 1% NaBH4, 50 mM DTT, or 100 mM β-mercaptoethanol. Has been used to immobilize proteins on supports containing amino groups. Chemical crosslinkers are used to covalently modify proteins for studying ligand receptor interactions, conformational changes in tertiary structure, or for protein labeling. Crosslinkers are divided into homobifunctional crosslinkers, containing two identical reactive groups, or heterobifunctional crosslinkers, containing two different reactive groups. Water-soluble cross-linking reagents help to avoid the use of organic solvents such as dimethylformamide (DMF) and dimethylsulfoxide (DMSO) when preparing bioconjugates.

Warning

Toxicity: Irritant (B)

Reconstitution

Following each use, purge the original vial with inert gas and refrigerate (4°C). Unstable in aqueous solution; reconstitute just prior to use. Alternatively, the solid DSP can be added directly to the sample to minimize hydrolysis. Hydrolysis is more pronounced in dilute protein solutions and can, therefore, be minimized by using more concentrated protein solutions (>5 mg/ml). For storage of stock solutions, reconstitute in anhydrous DMSO.

Other Notes

Howard, A.D., et al. 1985. J. Biol. Chem.260, 10833.
Lee, W.T., and Conrad, D.H. 1985. J. Immunol.134, 518.
D′Souza, S.E., et al. 1988. J. Biol. Chem.263, 3943.
Yamada, H., et al. 1981. Biochem.20, 4836.
Annunziato, M.E., et al. 1993. Bioconjugate Chem.4, 212.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service