Skip to Content
Merck
All Photos(1)

Key Documents

204870-M

Sigma-Aldrich

Complement C3d, Human

Complement C3d, Human is a native, 30 kDa polypeptide formed by the limited digestion of iC3b by trypsin or elastin. The binding of C3d to CR2 on B lymphocytes enhances B cell activation.

Synonym(s):

Complement Protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

Quality Level

Assay

≥85% (SDS-PAGE)

form

liquid

does not contain

preservative

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

impurities

IgG, IgM, IgA, C5, Factor H, Factor I, trace

storage temp.

−70°C

General description

Native, human C3d complement component. C3d is formed by the limited digestion of iC3b by trypsin or elastin. The C3d region of C3 contains the portion of the native C3 molecule which is capable of forming a covalent bond attachment to target surfaces under attack by complement. C3d is a polypeptide of M.W. 30 kDa. The binding of C3d to CR2 (CD21) receptors present on B lymphocytes markedly enhances B cell activation initiated by a wide variety of stimuli.

Warning

Toxicity: Standard Handling (A)

Physical form

In PBS, pH 7.2.

Preparation Note

Prepared from serum that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Luxembourg, A.T. and Cooper, N.R. 1994. J. Immunol.153, 4448.
Lambris, J.D., et al. 1985. Proc. Natl. Acad. Sci. USA82, 4235.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service