Skip to Content
Merck
All Photos(2)

Key Documents

1.94507

Supelco

Water

Distilled, EMPLURA®

Synonym(s):

Water

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2O
CAS Number:
Molecular Weight:
18.02
Beilstein:
2050024
MDL number:
UNSPSC Code:
12191602
bp:
100 °C (lit.)

Product Name

Water, (distilled), EMPLURA®

product line

EMPLURA®

form

(distilled)

refractive index

n20/D 1.34 (lit.)

bp

100 °C (lit.)

mp

0 °C (lit.)

density

1.000 g/mL at 3.98 °C (lit.)

storage temp.

room temp

SMILES string

O

InChI

1S/H2O/h1H2

InChI key

XLYOFNOQVPJJNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

1.94507.5021: 5 L Minimum Order Quantity: 4

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Inorganics and solvents for basic and standard applications manufactured locally.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service