Skip to Content
Merck
All Photos(1)

Documents

1.46763

Millipore

ReadyPlate 55 Plate Count Agar

for aerobic bacteria, suitable for filtration, diam. 55 mm, box of 20 plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106217
NACRES:
NA.74

Agency

ISO

Quality Level

form

plate (ready-to-use)

feature

ready-to-use

packaging

box of 20 plates

manufacturer/tradename

ReadyPlate

technique(s)

cell counting: suitable (total viable count (TVC))

diam.

55 mm

application(s)

filtration
food and beverages
microbiology

storage temp.

15-25°C

suitability

aerobic bacteria (total count)

General description

Application: Determination of total count of aerobic bacteria, Membrane filtration method

Packaging: 55 mm plate (20 plates per box)

Synonym: Casein-Peptone Dextrose Yeast Agar, Standard Methods Agar, PCA

Legal Information

READYPLATE is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

New microbiological standards for fruit juices focus on detecting Alicyclobacillus spp. using BAT broth and agar.

New microbiological standards for fruit juices focus on detecting Alicyclobacillus spp. using BAT broth and agar.

New microbiological standards for fruit juices focus on detecting Alicyclobacillus spp. using BAT broth and agar.

New microbiological standards for fruit juices focus on detecting Alicyclobacillus spp. using BAT broth and agar.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service