Skip to Content
Merck
All Photos(1)

Key Documents

1.14723

Supelco

Halogen lamp

for use with SPECTROQUANT® SQ 118

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115311

compatibility

for use with SPECTROQUANT® SQ 118

storage temp.

2-30°C

General description

SQ 118 Spectroquant®

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service