Skip to Content
Merck
All Photos(1)

Key Documents

110138

Millipore

N-Acetylneuraminic Acid, Synthetic

A component of mucoproteins, mucopolysaccharides, and mucolipids.

Synonym(s):

N-Acetylneuraminic Acid, Synthetic, NANA, Lactaminic Acid, Sialic Acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H19NO9
CAS Number:
Molecular Weight:
309.27
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.79
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

water: ≥10 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

N(C1C(OC(CC1O)(O)C(=O)O)C(O)C(O)CO)C(=O)C

InChI

1S/C11H19NO9/c1-4(14)12-7-5(15)2-11(20,10(18)19)21-9(7)8(17)6(16)3-13/h5-9,13,15-17,20H,2-3H2,1H3,(H,12,14)(H,18,19)

InChI key

SQVRNKJHWKZAKO-UHFFFAOYSA-N

General description

A component of mucoproteins, mucopolysaccharides, and mucolipids.

Warning

Toxicity: Standard Handling (A)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service