Skip to Content
Merck
All Photos(2)

Documents

1.09951

Supelco

Acetic acid

for 500 ml, c(CH₃COOH) = 1 mol/l (1 N) Titrisol®

Synonym(s):

Acetic acid solution, Methane carboxylic acid solution, Methylformic acid solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2H
CAS Number:
Molecular Weight:
60.05
Beilstein:
506007
MDL number:
UNSPSC Code:
12352106

product line

Titrisol®

form

liquid

expl. lim.

4-19.9 % (v/v) acetic acid)

reaction suitability

reaction type: Acid-base reactions

concentration

1 M

technique(s)

titration: suitable

pH

2.5 (20 °C, 20 g/L in H2O)

bp

102 °C

mp

-25 °C

transition temp

flash point >100 °C (External MSDS)

density

1.06 g/cm3 at 20 °C

SMILES string

CC(O)=O

InChI

1S/C2H4O2/c1-2(3)4/h1H3,(H,3,4)

InChI key

QTBSBXVTEAMEQO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Titrisol is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(C)

> 100 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service