Skip to Content
Merck
All Photos(2)

Key Documents

1.09259

Supelco

Water standard 5 mg/ml

(1 ml contains 5 mg H₂O) Aquastar®

Synonym(s):

Aquastar® Water Standard 5 mg/ml

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
EC Index Number:
200-752-1
NACRES:
NA.24
Pricing and availability is not currently available.

vapor pressure

2.04 hPa ( 20 °C)

Quality Level

product line

Aquastar®

form

liquid

autoignition temp.

300 °C

expl. lim.

1.6-8.0 % (v/v) 1-Pentanol)

technique(s)

Karl-Fischer titration: suitable
titration: suitable

bp

138 °C/1013 hPa

mp

-78 °C

transition temp

flash point 49 °C

density

0.82 g/cm3 at 20 °C

format

single component solution

storage temp.

15-25°C

General description

Water standard 5 mg/ml is a liquid water standard based on long-chain alcohol.

Application

The 5mg/ml water standard can be used as an analytical standard for daily titer control.

Analysis Note

Assay: 4.980 - 5.020 mg/ml

Legal Information

AQUASTAR is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

120.2 °F

Flash Point(C)

49 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service