Skip to Content
Merck
All Photos(1)

Key Documents

1.07872

Supelco

Strontium nitrate

for analysis EMSURE®

Synonym(s):

Strontium nitrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Sr(NO3)2
CAS Number:
Molecular Weight:
211.63
MDL number:
UNSPSC Code:
12352302
EC Index Number:
233-131-9
NACRES:
NA.21
Assay:
≥99.0% (complexometric)
form:
solid
solubility:
660 g/L
Pricing and availability is not currently available.

Quality Level

product line

EMSURE®

Assay

≥99.0% (complexometric)

form

solid

potency

2750 mg/kg LD50, oral (Rat)

pH

5-7 (20 °C, 50 g/L in H2O)

bp

645 °C/1013 hPa

mp

570 °C

solubility

660 g/L

density

2.986 g/cm3 at 20 °C

bulk density

1000 kg/m3

anion traces

chloride (Cl-): ≤0.001%

cation traces

Ba: ≤0.01%
Ca: ≤0.005%
Cu: ≤0.0002%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.005%
Na: ≤0.005%
Pb: ≤0.0002%

storage temp.

2-30°C

SMILES string

[Sr](O[N+](=O)[O-])O[N+](=O)[O-]

InChI

1S/2NO3.Sr/c2*2-1(3)4;/q2*-1;+2

InChI key

DHEQXMRUPNDRPG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Synthesis and Antimicrobial Analysis of High Surface Area Strontium-Substituted Calcium Phosphate Nanostructures for Bone Regeneration: The study explores the potential of strontium nitrate in bone regeneration through the synthesis of high surface area strontium-substituted calcium phosphate nanostructures with antimicrobial properties (Anwar et al., 2023).
  • Electrochemically Assisted Deposition of Strontium Modified Magnesium Phosphate on Titanium Surfaces: The article describes the use of strontium nitrate in creating coatings on titanium surfaces for improved biomedical applications, enhancing material properties through electrochemical deposition techniques (Meininger et al., 2016).

  • Production of Y-86 and Other Radiometals for Research Purposes Using a Solution Target System: This study involves the use of strontium nitrate in the production of radiometals, including Y-86, for various research and diagnostic purposes, employing a solution target system for effective synthesis (Oehlke et al., 2015).

  • Template-based Fabrication of SrTiO3 and BaTiO3 Nanotubes: The research discusses the utility of strontium nitrate in the template-based fabrication of nanotubes, specifically SrTiO3 and BaTiO3, which are critical in enhancing material properties for various industrial and technological applications (Chen et al., 2009).

Analysis Note

Assay (complexometric): ≥ 99.0 %
pH-value (5 %; water): 5.0 - 7.0
Chloride (Cl): ≤ 0.001 %
Ba (Barium): ≤ 0.01 %
Ca (Calcium): ≤ 0.005 %
Cu (Copper): ≤ 0.0002 %
Fe (Iron): ≤ 0.0005 %
K (Potassium): ≤ 0.005 %
Mg (Magnesium): ≤ 0.005 %
Na (Sodium): ≤ 0.005 %
Pb (Lead): ≤ 0.0002 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame over circleCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Ox. Sol. 1

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service