Skip to Content
Merck
All Photos(1)

Key Documents

1.07806

Supelco

Tin

granulated for analysis (particle size about 4 mm) EMSURE® Reag. Ph Eur

Synonym(s):

Tin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Sn
CAS Number:
Molecular Weight:
118.71
MDL number:
UNSPSC Code:
12141745
EC Index Number:
231-141-8
NACRES:
NA.21
form:
solid
Pricing and availability is not currently available.

Agency

reag. Ph. Eur.

Quality Level

product line

EMSURE®

form

solid

bp

2260 °C/1013 hPa

mp

232 °C

density

7.3 g/cm3 at 20 °C

cation traces

As: ≤0.001%
Bi: ≤0.004%
Cu: ≤0.001%
Fe: ≤0.005%
Pb: ≤0.005%
Sb: ≤0.02%
Zn: ≤0.002%

storage temp.

2-30°C

SMILES string

[Sn]

InChI

1S/Sn

InChI key

ATJFFYVFTNAWJD-UHFFFAOYSA-N

Related Categories

Application


  • Construction of biochar-based organohalide-respiring bacterial agent for remediation of 2,4,6-trichlorophenol contaminated soil: This study explores Triton X-100′s role in environmental remediation through the construction of a biochar-based bacterial agent, addressing soil contamination and highlighting its potential for broader applications in environmental science (Chen et al., 2024).

  • Development of porcine skeletal muscle extracellular matrix-derived hydrogels with improved properties and low immunogenicity: Triton X-100 was utilized in the development of biocompatible hydrogels from porcine skeletal muscle, underscoring its significance in biotechnological applications and medical materials science (Barajaa et al., 2024).

  • Formation of Alternating Surfactant-Enriched and Surfactant-Depleted Phases in the Taylor Cone of a Nanoelectrospray: This research details the phase behavior of surfactants like Triton X-100 in nanoelectrospray applications, relevant to analytical chemistry and material science fields (Han et al., 2024).

  • Study on active capacity and detergent application potential of low-temperature alkaline serine protease produced by new strain Exiguobacterium indicum 1.2.3: Investigates Triton X-100′s efficacy as a component of enzyme-based detergents, offering insights into its industrial applications in bioprocessing and manufacturing (Kati & Balci, 2023).

  • Enhanced crystalline cellulose degradation by a novel metagenome-derived cellulase enzyme: Discusses the role of Triton X-100 in enhancing the enzymatic breakdown of cellulose, pivotal for industrial biotechnology and renewable energy sectors (Kholousi Adab et al., 2024).

Analysis Note

As (Arsenic): ≤ 0.001 %
Bi (Bismuth): ≤ 0.004 %
Cu (Copper): ≤ 0.001 %
Fe (Iron): ≤ 0.005 %
Pb (Lead): ≤ 0.005 %
Sb (Antimony): ≤ 0.02 %
Zn (Zinc): ≤ 0.002 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service