Skip to Content
Merck
All Photos(1)

Key Documents

1.07297

Supelco

Phthalein purple

metal indicator, Reag. Ph Eur

Synonym(s):

Phthalein purple, Xylenolphthalein-bis(iminodiacetic acid), Metal phthaleine, Cresol phthalexone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H32N2O12
CAS Number:
Molecular Weight:
636.60
MDL number:
UNSPSC Code:
12171502
EC Index Number:
219-318-8
NACRES:
NA.21
Pricing and availability is not currently available.

Product Name

Phthalein purple, metal indicator Reag. Ph Eur

Agency

reag. Ph. Eur.

Quality Level

form

solid

impurities

≤10% Water (Karl Fischer)

mp

186 °C

bulk density

50 kg/m3

λmax

577-580 nm in 0.1 M NaOH

storage temp.

2-30°C

SMILES string

[Na+].[Na+].[Na+].[Na+].N(Cc1c(c(cc(c1)C3(OC(=O)c4c3cccc4)c2cc(c(c(c2)C)O)CN(CC(=O)[O-])CC(=O)[O-])C)O)(CC(=O)[O-])CC(=O)[O-]

InChI

1S/C32H32N2O12.4Na/c1-17-7-21(9-19(29(17)43)11-33(13-25(35)36)14-26(37)38)32(24-6-4-3-5-23(24)31(45)46-32)22-8-18(2)30(44)20(10-22)12-34(15-27(39)40)16-28(41)42;;;;/h3-10,43-44H,11-16H2,1-2H3,(H,35,36)(H,37,38)(H,39,40)(H,41,42);;;;/q;4*+1/p-4

InChI key

KSMPPVYJZJBIES-UHFFFAOYSA-J

Analysis Note

Identity (UV/VIS-Spectrum): passes test
Absorption maximum λmax.(Sodium hydroxide solution 0.1 mol/l): 577 - 580 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.005 g/l;sodium hydroxide solution 0.1 mol/l;calc. on anhydrous substance): ≥ 750
Water (according to Karl Fischer): ≤ 10 %
Suitability as indicator (for metal titration): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service