Skip to Content
Merck
All Photos(1)

Key Documents

1.05819

Supelco

Magnesium acetate tetrahydrate

for analysis EMSURE® ACS,Reag. Ph Eur

Synonym(s):

Magnesium acetate tetrahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3COO)2Mg · 4 H2O
CAS Number:
Molecular Weight:
214.45
MDL number:
UNSPSC Code:
12352107
EC Index Number:
205-554-9
NACRES:
NA.21
grade:
ACS reagent
form:
solid
solubility:
1200 g/L
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

product line

EMSURE®

Assay

99.5-102.0% (complexometric)

form

solid

impurities

≤0.005% Insoluble matter

pH

6.1 (20 °C, 50 g/L in H2O)

mp

80 °C

solubility

1200 g/L

density

1.45 g/cm3

bulk density

510 kg/m3

anion traces

chloride (Cl-): ≤0.001%
sulfate (SO42-): ≤0.001%

cation traces

Ba: ≤0.001%
Ca: ≤0.001%
Cu: ≤0.0005%
Fe: ≤0.0001%
K: ≤0.005%
Mn: ≤0.001%
Na: ≤0.001%
Sr: ≤0.005%
Zn: ≤0.0002%
heavy metals (as Pb): ≤0.0005%

storage temp.

2-30°C

SMILES string

[Mg+2].[O-]C(=O)C.[O-]C(=O)C.O.O.O.O

InChI

1S/2C2H4O2.Mg.4H2O/c2*1-2(3)4;;;;;/h2*1H3,(H,3,4);;4*1H2/q;;+2;;;;/p-2

InChI key

XKPKPGCRSHFTKM-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application


  • Optimisation of a sol-gel synthesis route for the preparation of MgF2 particles for a large scale coating process: Though focused on magnesium fluoride, this research could be analogous to processes involving magnesium acetate tetrahydrate, particularly in large-scale industrial applications related to material sciences, showcasing techniques for optimizing synthesis protocols for better industrial application and scalability (Scheurell et al., 2015).

  • Crystallization and preliminary crystallographic analysis of β-mannanase from Bacillus licheniformis: This article discusses the crystallization processes which might leverage magnesium acetate tetrahydrate as a crystallizing agent, showing potential applications in biochemical assay development and pharmaceutical biotechnology, which could be of interest to researchers in biochemistry and pharmaceutical fields (Songsiriritthigul et al., 2011).

Analysis Note

Assay (complexometric): 99.5 - 102.0 %
Insoluble matter: ≤ 0.005 %
Chloride (Cl): ≤ 0.001 %
Sulfate (SO₄): ≤ 0.001 %
Heavy metals (as Pb): ≤ 0.0005 %
Ba (Barium): ≤ 0.001 %
Ca (Calcium): ≤ 0.001 %
Cu (Copper): ≤ 0.0005 %
Fe (Iron): ≤ 0.0001 %
K (Potassium): ≤ 0.005 %
Mn (Manganese): ≤ 0.001 %
Na (Sodium): ≤ 0.001 %
Sr (Strontium): ≤ 0.005 %
Zn (Zinc): ≤ 0.0002 %
Corresponds to ACS,Reag. Ph Eur

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service