Skip to Content
Merck
All Photos(1)

Key Documents

1.04724

Supelco

Indigo carmine (C.I. 73015)

for analysis

Synonym(s):

Indigo carmine (C.I. 73015)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H8N2Na2O8S2
CAS Number:
Molecular Weight:
466.35
MDL number:
UNSPSC Code:
12171506
EC Index Number:
212-728-8
NACRES:
NB.21
Pricing and availability is not currently available.

Quality Level

potency

>2000 mg/kg LD50, oral (Rat)

composition

dye content, ≥80% spectrophotometric assay

loss

≤10% loss on drying, 110°C

pH

7 (20 °C, 10 g/L in H2O)

mp

>340 °C

solubility

1 g/L

density

0.71 g/cm3 at 29 °C

bulk density

700‑900 kg/m3

λmax

608-612 nm in water

storage temp.

2-30°C

SMILES string

[Na+].[Na+].[S](=O)(=O)([O-])c1cc2c(cc1)N\C(=C3\Nc4c(cc(cc4)[S](=O)(=O)[O-])C\3=O)\C2=O

InChI

1S/C16H10N2O8S2.2Na/c19-15-9-5-7(27(21,22)23)1-3-11(9)17-13(15)14-16(20)10-6-8(28(24,25)26)2-4-12(10)18-14;;/h1-6,17-18H,(H,21,22,23)(H,24,25,26);;/q;2*+1/p-2/b14-13+;;

InChI key

KHLVKKOJDHCJMG-QDBORUFSSA-L

Analysis Note

Dye content (spectrophotometrically): ≥ 80 %
Identity (UV/VIS-Spectrum): passes test
Absorption maximum λmax. (water): 608 - 612 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.01 g/l; water): 380 - 475
TLC-Test: passes test
Loss on drying (110 °C): ≤ 10 %

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service