Skip to Content
Merck
All Photos(1)

Key Documents

1.04633

Supelco

Hypophosphorous acid

50%, for analysis EMSURE®

Synonym(s):

Hypophosphorous acid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352301
NACRES:
NA.21

Product Name

Hypophosphorous acid 50%, for analysis EMSURE®

grade

reagent

Quality Level

vapor pressure

30 hPa ( 20 °C)

product line

EMSURE®

form

liquid

concentration

≥49.5% (alkalimetric)

impurities

≤0.0001% Cd (Cadmium)
≤0.0001% Zn (Zinc)
≤0.0005% As (Arsenic)
≤0.0005% Cu (Copper)
≤0.0005% Mn (Manganese)
≤0.0005% Ni (Nickel)
≤0.0005% Pb (Lead)
≤0.001% K (Potassium)
≤0.005% Ba (Barium)
≤0.005% Ca (Calcium)
≤0.005% Fe (Iron)
≤0.015% Chloride (Cl)
≤0.02% Total sulfur (as SO4)
≤0.10% Na (Sodium)

pH

1 (20 °C in H2O)

bp

108 °C/1013 hPa (decomposition)

mp

<-25 °C

density

1.21-1.26 g/cm3 at 20 °C

storage temp.

no temp limit

Application


  • One-Pot Synthesis and in Silico Molecular Docking Studies of Arylselanyl Hydrazides as Potential Antituberculosis Agents.: This research illustrates the utilization of 50% Hypophosphorous acid in the synthesis of arylselanyl hydrazides, emphasizing its role as a reducing agent to facilitate novel antituberculosis compounds, reflecting its significance in medicinal chemistry applications (Borges et al., 2022).

  • Formation of Thick Aminophenyl Films from Aminobenzenediazonium Ion in the Absence of a Reduction Source.: This study explores the action of 50% Hypophosphorous acid in the spontaneous formation of aminophenyl films from diazonium ions without additional reducing agents, highlighting its potential in surface coating processes (Simons et al., 2014).


Analysis Note

Assay (alkalimetric): ≥ 49.5 %
Phosphorous acid and phosphoric acid (as H₃PO₃): ≤ 2
Chloride (Cl): ≤ 0.015 %
Total sulfur (as SO₄): ≤ 0.02 %
As (Arsenic): ≤ 0.0005 %
Ba (Barium): ≤ 0.005 %
Ca (Calcium): ≤ 0.005 %
Cd (Cadmium): ≤ 0.0001 %
Cu (Copper): ≤ 0.0005 %
Fe (Iron): ≤ 0.005 %
K (Potassium): ≤ 0.001 %
Mn (Manganese): ≤ 0.0005 %
Na (Sodium): ≤ 0.10 %
Ni (Nickel): ≤ 0.0005 %
Pb (Lead): ≤ 0.0005 %
Zn (Zinc): ≤ 0.0001 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service