Skip to Content
Merck
All Photos(1)

Key Documents

100069

Sigma-Aldrich

17-DMAG

A potent antitumor analog of 17-AAG that binds to the ATPase site of human Hsp90α with high affinity and displays excellent bioavailability and aqueous solubility.

Synonym(s):

17-DMAG, NSC 707545, 17-Desmethoxy-17-N,N-dimethylaminoethylamino-geldanamycin, HCl, 17-N,N-Dimethylaminoethylamino-17-demethoxy-geldanamycin, HCl, 17DMAG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H48N4O8 · xHCl
CAS Number:
Molecular Weight:
616.75 (free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

purple

solubility

water: 10 mg/mL
DMSO: 5 mg/mL

shipped in

wet ice

storage temp.

−20°C

InChI

1S/C32H48N4O8.ClH/c1-18-14-22-27(34-12-13-36(5)6)24(37)17-23(29(22)39)35-31(40)19(2)10-9-11-25(42-7)30(44-32(33)41)21(4)16-20(3)28(38)26(15-18)43-8;/h9-11,16-18,20,25-26,28,30,34,38H,12-15H2,1-8H3,(H2,33,41)(H,35,40);1H/b11-9-,19-10+,21-16+;/t18-,20+,25+,26+,28-,30+;/m1./s1

InChI key

DFSYBWLNYPEFJK-IHLRWNDRSA-N

General description

A cell-permeable, water soluble, potent anti-tumor analog of 17-AAG (Cat. No. 100068) that binds to the ATPase site of human Hsp90α with high affinity (GI50 = 51 nM for 17-DMAG vs. 120 nM for 17-AAG in the NCI 60-cell panel in vitro activity screen).
A potent antitumor analog of 17-AAG (Cat. No. 100068) that binds to the ATPase site of human Hsp90α with high affinity (GI50 = 51 nM for 17-DMAG vs. 120 nM for 17-AAG in the NCI 60-cell panel in vitro activity screen), and displays excellent bioavailability and aqueous solubility.

Biochem/physiol Actions

Cell permeable: no
GI50 = 51 nM for 17-DMAG vs. 120 nM for 17-AAG in the NCI 60-cell panel in vitro activity screen
Primary Target
17-DMAG
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Tian, Z.Q., et al. 2004. Bioorg. Med. Chem.12, 5317.
Jez, J.M., et al. 2003. Chem. Biol.10, 361.
Egorin, M.J., et al. 2002. Cancer Chemother. Pharmacol.49, 7.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Behnoush Khaledian et al.
Cancer science, 112(3), 1225-1234 (2020-12-29)
We have previously identified receptor tyrosine kinase-like orphan receptor 1 (ROR1) as a direct transcriptional target of TTF-1/NKX2-1, a lineage-survival oncogene in lung adenocarcinoma. ROR1 sustains prosurvival signaling from multiple receptor tyrosine kinases including epidermal growth factor receptor, MET, and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service