Skip to Content
Merck
All Photos(2)

Key Documents

07-1817

Sigma-Aldrich

Anti-phospho-SRPK2 (Ser494) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Serine/threonine-protein kinase SRPK2, SFRS protein kinase 2, Serine/arginine-rich protein-specific kinase 2, SR-protein-specific kinase 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

species reactivity (predicted by homology)

Xenopus (based on 100% sequence homology), rat (based on 100% sequence homology)

technique(s)

inhibition assay: suitable (peptide)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer494)

Gene Information

human ... SRPK2(6733)

General description

Along with Serine/threonine protein kinase 2 (SRPK2), SRPK1 phosphorylates serine residues found in RS-domain-containing proteins, such as SFRS1 and SFRS2. These kinases also phosphorylate the same serine residues on the HBV core protein both in vitro and in vivo. The cytoplasmic proteins are critical for nuclear import of SR proteins in a phosphorylation-dependent manner. Additionally, they mediate the trafficking of splicing factors and play a role in spliceosome assembly and mitogenesis.

Specificity

This antibody is specific for the protein kinase domain of SRPK2 phosphorylated at Ser494, independent of observed phosphorylation at Ser497.

Immunogen

Epitope: Protein kinase domain
KLH-conjugated linear peptide corresponding to the protein kinase domain of SRPK2 phosphorylated at Ser494.

Application

Anti-phospho-SRPK2 (Ser494) Antibody is an antibody against phospho-SRPK2 (Ser494) for use in WB, PIA.
Peptide Inhibition Analysis: 1 Āµg/mL from a representative lot detected SRPK2 in 10 Āµg of insulin treated HepG2 cell lysate.
Research Category
Signaling
Research Sub Category
Cell Cycle, DNA Replication & Repair

Quality

Evaluated by Western Blot in untreated and insulin treated HepG2 cell lysate.

Western Blot Analysis: 1 Āµg/mL of this antibody detected SRPK2 in 10 Āµg HepG2 cell lysate.

Target description

~120 kDa observed. Two isoforms at ~77 kDa and ~79 kDa may be observed in some cell lysates.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8Ā°C from date of receipt.

Analysis Note

Control
Untreated and insulin treated HepG2 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product? Ā 

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a productā€™s label following the words ā€˜Lotā€™ or ā€˜Batchā€™.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mousumi Khatun et al.
Hepatology (Baltimore, Md.), 74(1), 41-54 (2020-11-26)
HCV often causes chronic infection in liver, cirrhosis, and, in some instances, HCC. HCV encodes several factors' those impair host genes for establishment of chronic infection. The long noncoding RNAs (lncRNAs) display diverse effects on biological regulations. However, their role
Aneesha Radhakrishnan et al.
Cancer biology & therapy, 17(2), 219-229 (2016-02-09)
Signaling plays an important role in regulating all cellular pathways. Altered signaling is one of the hallmarks of cancers. Phosphoproteomics enables interrogation of kinase mediated signaling pathways in biological systems. In cancers, this approach can be utilized to identify aberrantly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service