Skip to Content
Merck
All Photos(1)

Key Documents

05-850

Sigma-Aldrich

Anti-p68 Antibody, clone204

clone 204, Upstate®, from mouse

Synonym(s):

Anti-G17P1, Anti-HUMP68, Anti-p68

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

204, monoclonal

species reactivity

human, rat, mouse

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... DDX5(1655)

General description

Also known as DDX5, p68 is a DEAD-box ATPase and RNA splicing factor.

Specificity

p68

Immunogen

Purified T antigen-related protein from Ad2+ ND2 virus-infected HeLa cells.

Application

Detect p68 with Anti-p68 Antibody, clone204 (Mouse Monoclonal Antibody), that has been shown to work in WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

RNA Binding Protein (RBP)

Quality

Routinely evaluated by immunoblot of nuclear extracts from HeLa cells

Target description

68kDa

Physical form

Format: Purified
Protein G Purified

Storage and Stability

2 years at -20°C

Analysis Note

Control
Positive Antigen Control: Catalog #12-309, Hela cell nuclear extract. Add an equal volume of Laemmli reducing sample buffer to 10 μL of extract and boil for 5 minutes to reduce the preparation. Load 20 μg of reduced extract per lane for minigels.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raphaël Métivier et al.
Cell, 115(6), 751-763 (2003-12-17)
Transcriptional activation of a gene involves an orchestrated recruitment of components of the basal transcription machinery and intermediate factors, concomitant with an alteration in local chromatin structure generated by posttranslational modifications of histone tails and nucleosome remodeling. We provide here
Kengo Takeda et al.
International journal of molecular sciences, 25(7) (2024-04-13)
Chronic myeloid leukemia (CML) is induced by the expression of the fused tyrosine kinase BCR-ABL, which is caused by a chromosomal translocation. BCR-ABL inhibitors have been used to treat CML; however, the acquisition of resistance by CML cells during treatment
Zhi-Ren Liu
Molecular and cellular biology, 22(15), 5443-5450 (2002-07-09)
Modulation of the interaction between U1 snRNP and the 5' splice site (5'ss) is a key event that governs 5'ss recognition and spliceosome assembly. Using the methylene blue-mediated cross-linking method (Z. R. Liu, A. M. Wilkie, M. J. Clemens, and
M J Ford et al.
Nature, 332(6166), 736-738 (1988-04-21)
The p68 protein is a highly conserved nuclear antigen that is thought to be important in the regulation of cell growth and division. It is found in dividing cells of all mammals and amphibians tested, but not in quiescent cells.
Sumanta K Pal et al.
ACS omega, 4(2), 2472-2481 (2019-03-16)
Pyrvinium was identified as the first small molecule inhibitor of the androgen receptor (AR) DNA-binding domain (DBD). It was also among the first small molecules shown to directly inhibit the activity of AR splice variants (ARVs), which has important clinical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service