Key Documents
Z693529
2mag stainless-steel bath for MIXdrive
for use MIXdrive 6/15/60 and MIXdrive 1/1 eco/1 XS
Synonym(s):
laboratory stirrers, magnetic stir plates, magnetic stirrers, stirrers
About This Item
UNSPSC Code:
41103806
NACRES:
NB.72
packaging
pack of 1 ea
manufacturer/tradename
2mag 40950
inside W × D × H
277 mm × 537 mm × 175 mm
outside W × D × H
315 mm × 575 mm × 190 mm
General description
Durable, robust stainless steel bath, easy to clean, suitable for MIXdrive 6/15, fitting for immersion thermostats (heating/cooling), easy to clean, even weld and electro-polished, framed ridge formed as handle, exact positioning of the stirrer in the case of use the stirrer under the bath, stirring point mark inside the bath, left-/right stop for optimized thermostat positioning (circulation pump) for use with MIXdrive 6 / MIXdrive 15.
Required but not supplied: Z693502 or Z693510
Choose from one of the most recent versions:
Certificates of Analysis (COA)