Key Documents
Z564850
Sigma-Aldrich
Wrap-it tie installing tool
Economy version
About This Item
UNSPSC Code:
41121800
NACRES:
NB.35
availability
not available in EU
General description
Automatically adjusts wrap-it tie tension to desired position. Tension activated cutting blade trims off excess tie.
Choose from one of the most recent versions:
Certificates of Analysis (COA)