Z564737
Sleeve stopper septa
bottom I.D. × O.D. 12.7 mm × 23.7 mm, red
Synonym(s):
Rubber Septum
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
material
natural rubber
Joint
joint ST/NS24/40
feature
Use with narrow-mouth bottles: 64, 80 oz, and 1 gal jugs
bottom I.D. × O.D.
12.7 mm × 23.7 mm
color
red
Looking for similar products? Visit Product Comparison Guide
General description
Sleeve stopper septa are manufactured from pure natural rubber for excellent resealability when used for cannulation techniques or from FKM rubber for high chemical resistance. Natural rubber septa are available in red and white colors and a soft rubber formulation that contains no fillers or coloring agents for super tactility and easy manipulation.
Storage Note for Natural Rubber Septa:
Store septa in a closed bag out of sunlight to retard blooming, which is the formation of a whitish film on the surface of the rubber. Blooming is a natural process and does not affect performance. Remove visible bloom by washing or wiping off before use.
Storage Note for Natural Rubber Septa:
Store septa in a closed bag out of sunlight to retard blooming, which is the formation of a whitish film on the surface of the rubber. Blooming is a natural process and does not affect performance. Remove visible bloom by washing or wiping off before use.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service