Key Documents
Z513113
Sigma-Aldrich
Aldrich® non-corrosive lecture-bottle gas regulator
delivery pressure 0-100 psig, CGA Inlet size 170
About This Item
UNSPSC Code:
41100000
NACRES:
NB.51
Pricing and availability is not currently available.
delivery pressure measuring range
0-100 psig
CGA Inlet size
170
General description
  • Single-stage
  • Inlet: CGA 170
  • Outlet: 1/4 NPT Male
  • Max. inlet pressure: 3,000 psig
  • Temp. range: -40 to 140°F
  • Compact chrome-plated brass body
  • Lecture-bottle gas regulators include a PTFE sealing washer
  • Order spare PTFE sealing washers separately
Other Notes
product is not intended for medical use
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
1–2 of 2 Questions  
  1. What is the product number for the PTFE washer for the Lecture Bottle Gas Regulators?
    1 answer
    1. Lecture-bottle gas regulators include a PTFE sealing washer. Spare PTFE sealing washers can be ordered.Product Z146994, PTFE sealing washers, CGA Inlet size 180 can be used for Products Z148504, Aldrich® corrosive lecture-bottle gas regulator, 0-15 psig delivery pressure, CGA Inlet size 180 and Z146714, Aldrich® non-corrosive lecture-bottle gas regulator, 0-15 psig delivery pressure, CGA Inlet size 180.Product Z513105, PTFE sealing washers, CGA Inlet size 170 can be used for Products Z512958, Aldrich® non-corrosive lecture-bottle gas regulator, 0-15 psig delivery pressure, CGA Inlet size 170 and Z513113, Aldrich® non-corrosive lecture-bottle gas regulator, 0-100 psig delivery pressure, CGA Inlet size 170.
      Helpful?
  2. What is the Department of Transportation shipping information for this product?
    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
      Helpful?