Key Documents
Z506567
Perfektum® three-way stainless steel stopcock
male Luer lock to female Luer to side female Luer
About This Item
UNSPSC Code:
41122000
NACRES:
NB.03
Pricing and availability is not currently available.
manufacturer/tradename
Cadence Science 6011SS
General description
Made from corrosion-resistant 303 stainless steel.
Legal Information
Perfektum is a registered trademark of Cadence Science
Choose from one of the most recent versions:
Certificates of Analysis (COA)