Skip to Content
Merck
All Photos(1)

Key Documents

Z407178

Sigma-Aldrich

Sigma-Aldrich® SDS Subscription

custom and intranet version, quarterly subscription

Synonym(s):

Safety Data Sheet Subscription

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
NACRES:
NE.24

Looking for similar products? Visit Product Comparison Guide

General description

Sigma-Aldrich Safety Data Sheets
Contains more than 300,000 Sigma-Aldrich chemicals
Features:
  • Emergency overview
  • HMIS and NFPA ratings
  • Transportation data from DOT, IATA, and ADR/AID
  • Canadian DSL & NDSL lists
Country specific content:
  • European country specific OELs
  • Swiss poison classes
  • Annex listings updated
  • And more...
SDS′s are customized with country-specific regulatory requirements, data and language.
Subscribers receive quarterly updates containing a complete database including new products and updated safety information. Quarterly updates may ship on DVD discs or USB Flash Drives. Search fields include: catalog number, CAS registry number; and more than 600,000 chemical names and synonyms.

Compatible with Windows XP, VISTA, and Windows 7*, and Windows 8*. Subscribers receive quarterly updates. Network versions are licensed on a concurrent user and server basis and require a signed license agreement. For information on network versions please contact your local sales office. Custom version includes data and indices appropriate for each file format.
*compatible with Windows 7 if produced after June 2011.

One year subscription.

Legal Information

SIGMA-ALDRICH is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service