Key Documents
Z291846
Supelco
Closures for WISP 48-position autosampler vials
screw-thread caps, open-top closures, black PP
About This Item
UNSPSC Code:
41122001
NACRES:
NB.11
material
black polypropylene cap
feature
closure type black polypropylene hole cap
packaging
pkg of 100 ea
fitting
thread for 13-425
Legal Information
WISP is a trademark of Waters